MilliporeSigma
  • Pharmacokinetics of 17 beta-dihydroequilin sulfate and 17 beta-dihydroequilin in normal postmenopausal women.
Pharmacokinetics of 17 beta-dihydroequilin sulfate and 17 beta-dihydroequilin in normal postmenopausal women.
The Journal of clinical endocrinology and metabolism (1994-01-01)
B R Bhavnani, A Cecutti
ABSTRACT
The MCRs of 17 beta-dihydroequilin sulfate and 17 beta-dihydroequilin were determined in normal postmenopausal women by single iv injection of either 17 beta-[3H]dihydroequilin sulfate ([3H]17 beta-EqS) or 17 beta-[3H]dihydroequilin ([3H]17 beta-Eq). After the administration of [3H]17 beta-EqS, blood was drawn at various time intervals, and the plasma obtained was fractionated into the unconjugated, sulfate, and glucuronide fractions. The bulk of radioactivity was present in the sulfate fraction, and from this [3H]17 beta-EqS, [3H]equilin sulfate, [3H]equilenin sulfate, and 17 beta-[3H]dihydroequilenin sulfate were isolated and purified, and their concentrations were measured. The disappearance of [3H]17 beta-EqS from plasma can be described as a function of two exponentials. The half-life of the initial fast component was 5 +/- 0.2 min; this component represents the distribution and transfer from a space, with a mean volume (V1) of 6 +/- 0.5 L. The value for the rate constant (k) of total removal from this space was 300 +/- 20 U/day, of which 35 +/- 2% was irreversible. The mean half-life of the slower component of 17 beta-EqS was 147 +/- 15 min, and the mean MCR was 376 +/- 93 L/day.m2. Similarly, after the administration of [3H]17 beta-Eq, the disappearance of radioactivity as 17 beta-Eq from plasma also had two components. The half-lives of the fast and slow component were 5.5 +/- 0.8 and 45 +/- 2.0 min, respectively. The MCR of 17 beta-Eq was 1252 +/- 103 L/day.m2. From both series of experiments, unconjugated and sulfate-conjugated equilin, equilenin, and 17 beta-dihydroequilenin were isolated and purified, and their concentrations were measured. No 17 alpha-reduced metabolites were detected. These results indicate that 17 beta-EqS is cleared twice as fast as equilin sulfate (MCR, 176 L/day.m2), whereas the more potent estrogen 17 beta-Eq is cleared 2 times slower than equilin. The slower elimination and greater estrogenic activity of 17 beta-Eq support the hypothesis that the major in vivo activity of equilin sulfate present in conjugated equine estrogen preparations is expressed via its metabolites 17 beta-EqS and 17 beta-Eq.
Product Description
17α-Dihydroequilin, European Pharmacopoeia (EP) Reference Standard
USP
17α-Dihydroequilin, United States Pharmacopeia (USP) Reference Standard