MilliporeSigma
  • Four polymorphisms of the CAPN 10 gene and their relationship to polycystic ovary syndrome susceptibility: a meta-analysis.
Four polymorphisms of the CAPN 10 gene and their relationship to polycystic ovary syndrome susceptibility: a meta-analysis.
Clinical endocrinology (2011-09-13)
Miaoling Huang, Jianpeng Xiao, Xiaomiao Zhao, Changhao Liu, Qing Chen
ABSTRACT
To investigate the association between CAPN 10 gene polymorphism and polycystic ovary syndrome (PCOS) susceptibility. Meta-analysis of published case-control studies of four single nucleotide polymorphisms (SNPs) in CAPN 10 and PCOS susceptibility. Women with PCOS. Odds ratios (ORs) and 95% confidence intervals (CIs) for heterozygous, homozygous, dominant model, recessive model and allele. A total of 11 studies were involved in the meta-analysis. UCSNP-63 was significantly associated with PCOS, with homozygous carriers (TT vs CC: OR = 0·64; 95% CI: 0·45-0·90) and recessive model (TT vs CC and CT: OR = 0·64; 95% CI: 0·45-0·90) being protective factors. In addition, UCSNP-19 was significantly associated with PCOS, with recessive model (ins/ins vs del/del and del/ins: OR = 0·72, 95% CI: 0·59-0·88) and insert allele (ins vs del: OR = 0·85, 95% CI: 0·76-0·96) being protective factors, while heterozygous carriers (del/ins vs del/del: OR = 1·56, 95% CI: 1·24-1·94) and deletion allele (del vs ins: OR = 1·18, 95% CI: 1·04-1·32) being risk factors. However, no significant associations were found between UCSNP-44, -43 and PCOS. Moreover, the results of the Rotterdam criteria subgroup analysis were similar with that of overall analysis. This is the first report on the association between CAPN 10 UCSNP-63 and PCOS in genotype, with homozygous carriers and recessive model being protective factors. Additionally, insert allele and recessive model of UCSNP-19 are protective factors, while deletion allele and heterozygous genotype are risk factors for PCOS development.