MilliporeSigma
  • Expression, purification and characterization of recombinant human interleukin-2-serum albumin (rhIL-2-HSA) fusion protein in Pichia pastoris.
Expression, purification and characterization of recombinant human interleukin-2-serum albumin (rhIL-2-HSA) fusion protein in Pichia pastoris.
Protein expression and purification (2012-05-23)
Jianyong Lei, Bo Guan, Bo Li, Zuoying Duan, Yun Chen, Huazhong Li, Jian Jin
ABSTRACT
Interleukin-2 (IL-2) plays important roles in variety of immune functions. Recombinant IL-2 has become an important therapeutic protein for therapy of melanoma and renal cell carcinoma. Previously, it was proved that the therapeutic efficacy of rIL-2 expressed in Saccharomyces cerevisiae was improved by prolonging its in vivo half-life through genetic fusion with albumin. In this study, a fusion protein composed of hIL-2 genetically fused to HSA was expressed as a secretory protein under AOX1 (alcohol oxidase 1) promoter in Pichia pastoris. An effective strategy was established to express rhIL-2-HSA fusion protein in 5L scale and the optimal purification procedure was investigated. The purity of rhIL-2-HSA in final product was about 95%. The purified rhIL-2-HSA fusion protein could be recognized by both anti-hIL-2 and anti-human serum albumin monoclonal antibody. Bioactivity analysis showed that the purified rhIL-2-HSA fusion protein displayed high level activity on proliferation in IL-2 dependent manner in CTLL2-cells. rhIL-2-HSA fusion protein also showed a extended half-life in plasma compared with IL-2 when tested in a BALB/c mouse model. This study provides an alternative strategy for large-scale production of bioactive rhIL-2-HSA fusion protein using P. pastoris as an expression host.