MilliporeSigma
Mometasone furoate/formoterol in the treatment of persistent asthma.
Expert review of respiratory medicine (2011-11-16)
William E Berger
ABSTRACT
Mometasone furoate and formoterol fumarate dihydrate (MF/F) administered via metered-dose inhaler with a dose counter is a new fixed-dose combination of an inhaled corticosteroid and a long-acting β2-agonist indicated for daily maintenance therapy in patients aged ≥12 years with persistent asthma. Randomized, controlled trials have suggested that MF/F reduces asthma deteriorations while improving lung function and other measures of asthma control, including quality-of-life. Clinical safety studies lasting up to 1 year have found that MF/F has a low incidence of local and systemic side effects.
Sigma-Aldrich
Mometasone furoate, ≥98% (HPLC)
Mometasone furoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Mometasone Furoate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mometasone furoate, United States Pharmacopeia (USP) Reference Standard