MilliporeSigma
  • Trimebutine: mechanism of action, effects on gastrointestinal function and clinical results.
Trimebutine: mechanism of action, effects on gastrointestinal function and clinical results.
The Journal of international medical research (1997-11-19)
M Delvaux, D Wingate
ABSTRACT
The actions of trimebutine [3,4,5-trimethoxybenzoic acid 2-(dimethylamino)-2-phenylbutylester] on the gastrointestinal tract are mediated via (i) an agonist effect on peripheral mu, kappa and delta opiate receptors and (ii) release of gastrointestinal peptides such as motilin and modulation of the release of other peptides, including vasoactive intestinal peptide, gastrin and glucagon. Trimebutine accelerates gastric emptying, induces premature phase III of the migrating motor complex in the intestine and modulates the contractile activity of the colon. Recently, trimebutine has also been shown to decrease reflexes induced by distension of the gut lumen in animals and it may therefore modulate visceral sensitivity. Clinically, trimebutine has proved to be effective in the treatment of both acute and chronic abdominal pain in patients with functional bowel disorders, especially irritable bowel syndrome, at doses ranging from 300 to 600 mg/day. It is also effective in children presenting with abdominal pain.