MilliporeSigma
  • Interactions between fluorinated cationic guar gum and surfactants in the dilute and semi-dilute solutions.
Interactions between fluorinated cationic guar gum and surfactants in the dilute and semi-dilute solutions.
Carbohydrate polymers (2013-11-28)
Chen Wang, Xiaorui Li, Peizhi Li, Yuhua Niu
ABSTRACT
The interactions between the fluorinated cationic guar gum (FCGG) and ionic surfactants including cetyl trimethyl ammonium bromide (CTAB) and sodium lauryl sulfate (SDS) were studied by light scattering, fluorescence spectroscopy, UV-spectrophotometer, (19)F NMR and dynamic rheometer, respectively. The FCGG is prepared with cationic guar gum, isophorone diisocyanate and 2,2,3,4,4,4-hexafluoro-1-butanol. The results show that, with the addition of the surfactants, the stretching degree of the FCGG chains is increased in the FCGG/CTAB solutions, while the dramatical shrinking of FCGG chain, the phase separation and the re-stretched macromolecules appear successively because of the electricity neutralization reaction in the FCGG/SDS system. The mixed hydrophobic domains in all solutions will be reinforced and then dismantled. The solution elasticity shows up the maximum value accordingly. The surfactants can be embedded in the micro-domains and then hinder the fluorinated segmental motions. The interactions between FCGG and SDS are much stronger than those between FCGG and CTAB.