MilliporeSigma
  • Quantitative study of natural antioxidant systems for cellular nitrofurantoin toxicity.
Quantitative study of natural antioxidant systems for cellular nitrofurantoin toxicity.
Biochimica et biophysica acta (1988-12-15)
C Michiels, J Remacle
ABSTRACT
The toxicity of nitrofurantoin was studied on human WI-38 fibroblasts: this chemical was lethal when added at concentrations higher than 5.10(-5) M in the culture medium. The protection afforded by antioxidants was then tested: alpha-tocopherol gave at 10(-4) M a light protection in contrast to ascorbic acid which even became toxic at high concentrations. We also tested catalase, superoxide dismutase and glutathione peroxidase introduced intracellularly by the microinjection technique. On a molecular basis, glutathione peroxidase was 23-times more efficient than catalase and 3000-times more than superoxide dismutase. The results also showed that a similar range of enzyme concentrations was found for the protection against high oxygen pressure. This suggests that, in the case of both oxygen and nitrofurantoin toxicity, the peroxide derivatives are the most toxic intermediates of the free radical attacks.
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications