MilliporeSigma
  • The interferon-inducible IFI16 gene inhibits tube morphogenesis and proliferation of primary, but not HPV16 E6/E7-immortalized human endothelial cells.
The interferon-inducible IFI16 gene inhibits tube morphogenesis and proliferation of primary, but not HPV16 E6/E7-immortalized human endothelial cells.
Experimental cell research (2004-01-20)
Ravera Raffaella, Daniela Gioia, Marco De Andrea, Paola Cappello, Mirella Giovarelli, Peggy Marconi, Roberto Manservigi, Marisa Gariglio, Santo Landolfo
ABSTRACT
Immunohistochemical analysis has demonstrated that the human IFI16 gene, in addition to the hematopoietic tissues, is highly expressed in endothelial cells and squamous stratified epithelia. In this study, we have developed a reliable HSV-derived replication-defective vector (TO-IFI16) to efficiently transduce IFI16 into primary human umbilical vein endothelial cells (HUVEC), which are usually poorly transfectable. HUVEC infection with TO-IFI16 virus suppressed endothelial migration, invasion and formation of capillary-like structures in vitro. In parallel, sustained IFI16 expression inhibited HUVEC cell cycle progression, accompanied by significant induction of p53, p21, and hypophosphorylated pRb. Further support for the involvement of these pathways in IFI16 activity came from the finding that infection with TO-IFI16 virus does not impair the in vitro angiogenic activity and cell cycle progression of HUVEC immortalized by HPV16 E6/E7 oncogenes, which are known to inactivate both p53 and pRb systems. This use of a reliable viral system for gene delivery into primary human endothelial cells assigns a potent angiostatic activity to an IFN-inducible gene, namely IFI16, and thus throws further light on antiangiogenic therapy employing IFNs.