MilliporeSigma
  • The human and murine protocadherin-beta one-exon gene families show high evolutionary conservation, despite the difference in gene number.
The human and murine protocadherin-beta one-exon gene families show high evolutionary conservation, despite the difference in gene number.
FEBS letters (2001-04-27)
K Vanhalst, P Kools, E Vanden Eynde, F van Roy
ABSTRACT
Extensive cDNA analysis demonstrated that all human and mouse protocadherin-beta genes are one-exon genes. The protein sequences of these genes are highly conserved, especially the three most membrane-proximal extracellular domains. Phylogenetic analysis suggested that this unique gene family evolved by duplication of one single protocadherin-beta gene to 15 copies. The final difference in the number of protocadherin-beta genes in man (#19) and mouse (#22) is probably caused by duplications later in evolution. The complex relationship between human and mouse genes and the lack of pseudogenes in the mouse protocadherin-beta gene cluster suggest a species-specific evolutionary pressure for maintenance of numerous protocadherin-beta genes.