MilliporeSigma
  • An electrochemical immunosensor for ultrasensitive detection of carbohydrate antigen 199 based on Au@Cu(x)OS yolk-shell nanostructures with porous shells as labels.
An electrochemical immunosensor for ultrasensitive detection of carbohydrate antigen 199 based on Au@Cu(x)OS yolk-shell nanostructures with porous shells as labels.
Biosensors & bioelectronics (2014-07-25)
Aiping Guo, Yueyun Li, Wei Cao, Xianchao Meng, Dan Wu, Qin Wei, Bin Du
ABSTRACT
A novel and sensitive electrochemical immunosensor for ultrasensitive detection of pancreatic cancer biomarker carbohydrate antigen 199 (CA199) was proposed by using Au@Cu(x)OS yolk-shell nanostructures with porous shells as labels for signal amplification. Au@Cu(x)OS yolk-shell nanostructures exhibit high electrocatalytic activity toward the reduction of hydrogen peroxide (H2O2) as analytical signal. Moreover, secondary antibody (Ab2) can adsorb on the surface of Au@Cu(x)OS with porous shells which has large surface area and could greatly increase the probability of Ab2-antigen interactions thereby leading to higher sensitivity. Reduced graphene oxide-tetraethylene pentamine (rGO-TEPA), containing abundant amine groups, was supported Au nanoparticles as a support platform to immobilize the primary antibody (Ab1). The resulting sensing interface of rGO-TEPA/AuNPs could provide a large electroconductive surface area, allowing high loadings of the biological recognition elements as well as the occurrence of electrocatalytic and electron-transfer processes. Under optimal conditions, the immunosensor exhibited a wide linear response to CA199 ranging from 0.001 to 12 U/mL with a low detection limit of 0.0005 U/mL. The designed immunosensor displayed good precision, high sensitivity, acceptable stability and reproducibility, and has been applied to the analysis of serum with satisfactory results. The proposed method provides a new promising platform of clinical immunoassay for other biomolecules.