MilliporeSigma
  • Isotope Effect on Dissociation Probabilities in the Mass Spectra of Chloroform and Chloroform-d.
Isotope Effect on Dissociation Probabilities in the Mass Spectra of Chloroform and Chloroform-<I>d</I>.
J. Chem. Phys. (1951)
Dibeler VH and Bernstein RB.