MilliporeSigma
  • Cocrystal screening of hydroxybenzamides with benzoic acid derivatives: a comparative study of thermal and solution-based methods.
Cocrystal screening of hydroxybenzamides with benzoic acid derivatives: a comparative study of thermal and solution-based methods.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-09-15)
Alex N Manin, Alexander P Voronin, Ksenia V Drozd, Nikolay G Manin, Annette Bauer-Brandl, German L Perlovich
ABSTRACT
The main problem occurring at the early stages of cocrystal search is the choice of an effective screening technique. Among the most popular techniques of obtaining cocrystals are crystallization from solution, crystallization from melt and solvent-drop grinding. This paper represents a comparative analysis of the following screening techniques: DSC cocrystal screening method, thermal microscopy and saturation temperature method. The efficiency of different techniques of cocrystal screening was checked in 18 systems. Benzamide and benzoic acid derivatives were chosen as model systems due to their ability to form acid-amide supramolecular heterosynthon. The screening has confirmed the formation of 6 new cocrystals. The screening by the saturation temperature method has the highest screen-out rate but the smallest range of application. DSC screening has a satisfactory accuracy and allows screening over a short time. Thermal microscopy is most efficient as an additional technique used to interpret ambiguous DSC screening results. The study also included an analysis of the influence of solvent type and component solubility on cocrystal formation.
Sigma-Aldrich
4-Hydroxybenzamide, 98%