MilliporeSigma
  • A Newly-Identified Polymorphism in Rhesus Macaque Complement Factor H Modulates Binding Affinity for Meningococcal FHbp.
A Newly-Identified Polymorphism in Rhesus Macaque Complement Factor H Modulates Binding Affinity for Meningococcal FHbp.
PloS one (2015-08-19)
Monica Konar, Peter T Beernink, Dan M Granoff
ABSTRACT
Two meningococcal serogroup B vaccines contain Factor H binding protein (FHbp). Binding of Factor H (FH) to FHbp was thought to be specific for human or chimpanzee FH. However, in a previous study an amino acid polymorphism in rhesus macaque FH domain 6, tyrosine at position 352 (Y352) was associated with high binding to FHbp, whereas histidine at position 352 (H352) was associated with low binding. Here we report that a second FH polymorphism at position 360 also affects macaque FH binding. Of 43 macaques, 11 had high FH binding and 32 had low binding. As in our previous study, all 11 animals with high binding had Y352, and 24 with low binding had H352. However the remaining eight with low FH binding had Y352, which was predicted to yield high binding. All eight had S360 instead of P360. Thus, three allelic variants at positions 352 and 360 affect macaque FH binding to FHbp: HP (low), YS (low), and YP (high). We measured binding affinity of each FH sequence type to FHbp by surface plasmon resonance. Two animals with high binding types (YS/YP and HP/YP) had dissociation constants (KD) of 10.4 and 18.2 nM, respectively, which were similar to human FH (19.8 nM). Two macaques with low binding (HP/HP and HP/YS) had KD values approximately five-fold higher (100.3 and 99.5 nM, respectively). A third macaque with low binding (YS/YS) had a KD value too high to be measured. Macaques have at least three allelic variants encoding FH with different affinities for FHbp (five genotypic combinations of these variants). Since in previous studies binding of FH to FHbp vaccines decreased protective antibody responses, our data will aid in selection of macaques with FH binding that is similar to humans for further investigation of FHbp vaccine immunogenicity.