MilliporeSigma
  • Pyridazines. Part XXIX: synthesis and platelet aggregation inhibition activity of 5-substituted-6-phenyl-3(2H)-pyridazinones. Novel aspects of their biological actions.
Pyridazines. Part XXIX: synthesis and platelet aggregation inhibition activity of 5-substituted-6-phenyl-3(2H)-pyridazinones. Novel aspects of their biological actions.
Bioorganic & medicinal chemistry (2002-07-12)
Eddy Sotelo, Nuria Fraiz, Matilde Yáñez, Vicente Terrades, Reyes Laguna, Ernesto Cano, Enrique Raviña
ABSTRACT
A series of 6-phenyl-3(2H)-pyridazinones with a diverse range of substituents in the 5-position have been prepared and evaluated in the search for new antiplatelet agents. A significant dependence of the substituent on the inhibitory effect has been observed. The pharmacological study of these compounds confirms that modification of the chemical group at position 5 of the 6-phenyl-3(2H)-pyridazinone system influences both variations in the antiplatelet activity and the mechanism of action.