MilliporeSigma
  • The effect of sodium alkyl sulfates (C8-C16) on the electrokinetic properties of hematite.
The effect of sodium alkyl sulfates (C8-C16) on the electrokinetic properties of hematite.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects (2016)
Kosmulski M and Maczka E.