MilliporeSigma
  • Regioselective enzymatic glycosylation of natural polyhydroxylated compounds: galactosylation and glucosylation of protopanaxatriol ginsenosides.
Regioselective enzymatic glycosylation of natural polyhydroxylated compounds: galactosylation and glucosylation of protopanaxatriol ginsenosides.
The Journal of organic chemistry (2001-06-30)
B Danieli, L Falcone, D Monti, S Riva, S Gebhardt, M Schubert-Zsilavecz
ABSTRACT
Ginsenoside Rg1 (1), the most representative Ginsenoside from Panax Ginseng C.A. Meyer belonging to the protopanaxatriol family, has been galactosylated by action of the beta-(1,4)-galactosyltransferase (GalT) from bovine colostrum, using UDP-galactose as an activated sugar donor. The enzyme showed the well-known specificity for the formation of a beta-linkage with the C-4 OH of the glucose acceptor, but it was not able to discriminate between the two glucose moieties of 1, giving a mixture of mono- and digalactosylated derivatives. Other natural Rg1-analogues such as F1, Rh1, Re, as well as the synthetic derivative 6'-O-acetyl-Rg1 have been also galactosylated, giving monolactosyl derivatives. GalT was also able to accept UDP-glucose as an activated sugar donor, giving rise to cellobiosyl derivatives of Rg1.