MilliporeSigma
  • Protection of neonatal mice from group B streptococcal infection by maternal immunization with beta C protein.
Protection of neonatal mice from group B streptococcal infection by maternal immunization with beta C protein.
Infection and immunity (1992-12-01)
L C Madoff, J L Michel, E W Gong, A K Rodewald, D L Kasper
ABSTRACT
Group B streptococci (GBS) cause the majority of cases of neonatal sepsis and meningitis in the United States. Immunization of women of childbearing age is one strategy under consideration for the prevention of neonatal disease. The beta C protein, a 130-kDa antigen present in many clinical isolates of GBS, was purified from GBS by extraction into sodium dodecyl sulfate (SDS)-containing buffer, preparative SDS-polyacrylamide gel electrophoresis, and electroelution. Purified beta C protein antigen (25 micrograms) with Freund's adjuvant was used to immunize rabbits. Rabbits developed enzyme-linked immunosorbent assay titers of > 1:1.6 x 10(6), and sera from immunized rabbits were administered to pregnant mice. Their neonatal pups were then challenged with a strain of GBS expressing beta C protein; 68% of these pups were protected by immune antiserum, whereas no controls were protected (P < 0.001). The immune serum (diluted 1:100) facilitated opsonophagocytic killing of GBS strains expressing the beta C protein but not those that do not express the antigen (mean log kill +/- standard deviation = 0.71 +/- 0.8 log10 CFU for beta+ strains and 0.09 +/- 0.2 for beta- strains; P = 0.02). In subsequent experiments, adult female mice were actively immunized with two doses of 2, 5, or 10 micrograms of beta C protein 2 months prior to mating. One- to two-day-old offspring of these dams were challenged with GBS and were protected in a dose-dependent manner, with 96% survival in the high-dose (10-micrograms) group and 20% survival in a sham-immunized control group (P < 0.001). Thus, active immunization of mice with the GBS beta C protein confers protection against lethal infection with beta+ GBS to their offspring.
Sigma-Aldrich
2,2′-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 99%
Pricing and availability is not currently available.