MilliporeSigma
  • New small nuclear RNA gene-like transcriptional units as sources of regulatory transcripts.
New small nuclear RNA gene-like transcriptional units as sources of regulatory transcripts.
PLoS genetics (2007-02-06)
Aldo Pagano, Manuele Castelnuovo, Federico Tortelli, Roberto Ferrari, Giorgio Dieci, Ranieri Cancedda
ABSTRACT
By means of a computer search for upstream promoter elements (distal sequence element and proximal sequence element) typical of small nuclear RNA genes, we have identified in the human genome a number of previously unrecognized, putative transcription units whose predicted products are novel noncoding RNAs with homology to protein-coding genes. By elucidating the function of one of them, we provide evidence for the existence of a sense/antisense-based gene-regulation network where part of the polymerase III transcriptome could control its polymerase II counterpart.