MilliporeSigma
  • Clinical significance of IL-2 and IL-10 gene polymorphisms and serum levels in patients with basal-cell carcinoma.
Clinical significance of IL-2 and IL-10 gene polymorphisms and serum levels in patients with basal-cell carcinoma.
Biomarkers in Medicine (2016)
Sobjanek M, et al.