MilliporeSigma
  • Investigation on the association between inerleukin-10 -592C/A, 819C/T and -1082A/G gene polymorphisms and development of diabetic nephrophathy.
Investigation on the association between inerleukin-10 -592C/A, 819C/T and -1082A/G gene polymorphisms and development of diabetic nephrophathy.
International Journal of Clinical and Experimental Pathology (2015)
Yin Q, et al.