MilliporeSigma
  • Calcyclin-binding protein inhibits proliferation, tumorigenicity, and invasion of gastric cancer.
Calcyclin-binding protein inhibits proliferation, tumorigenicity, and invasion of gastric cancer.
Molecular cancer research : MCR (2008-01-04)
Xiaoxuan Ning, Shiren Sun, Liu Hong, Jie Liang, Lili Liu, Shuang Han, Zhiguo Liu, Yongquan Shi, Yuan Li, Weiqin Gong, Shanhong Zhang, Yu Chen, Xueyan Guo, Yi Cheng, Kaichun Wu, Daiming Fan
ABSTRACT
Calcyclin-binding protein/Siah-1-interacting protein (CacyBP/SIP), a target protein of the S100 family, which includes S100A6, S100A1, S100A12, S100B, and S100P, has been identified as a component of a novel ubiquitinylation complex leading to beta-catenin degradation. However, the function of CacyBP/SIP in gastric cancer has not been elucidated. In the present study, we prepared CacyBP/SIP overexpressing and knockdown cell lines of gastric cancer. Forced CacyBP/SIP expression inhibited the proliferation of gastric cancer cells, suppressed tumorigenicity in vitro, and prolonged the survival time of tumor-bearing nude mice. In addition, increased CacyBP/SIP repressed the invasive potential of gastric cancer cells. Conversely, the down-regulation of CacyBP/SIP by RNA interference showed the opposite effects. Further studies showed that depressed CacyBP/SIP increased the expression of total and nuclear beta-catenin at the protein level and elevated the transcriptional activity of Tcf/LEF. Taken together, our results suggest that CacyBP/SIP may be a potential inhibitor of cell growth and invasion in the gastric cancer cell, at least in part through the effect on beta-catenin protein expression and transcriptional activation of Tcf/LEF.