MilliporeSigma
Closing in on the biological functions of Fps/Fes and Fer.
Nature reviews. Molecular cell biology (2002-05-08)
Peter Greer
ABSTRACT
Fps/Fes and Fer are the only known members of a distinct subfamily of the non-receptor protein-tyrosine kinase family. Recent studies indicate that these kinases have roles in regulating cytoskeletal rearrangements and inside out signalling that accompany receptor ligand, cell matrix and cell cell interactions. Genetic analysis using transgenic mouse models also implicates these kinases in the regulation of inflammation and innate immunity.