MilliporeSigma
  • Facile Enantioselective Ring-Opening Reaction of meso Epoxides with Anilines Using (S)-(-)-BINOL-Ti Complex as a Catalyst.
Facile Enantioselective Ring-Opening Reaction of meso Epoxides with Anilines Using (S)-(-)-BINOL-Ti Complex as a Catalyst.
European Journal of Organic Chemistry (2006)
Kureshy RI, et al.