MilliporeSigma
  • Highly Enantioselective Aryl Additions of [AlAr3 (thf)] to Ketones Catalyzed by a Titanium (IV) Catalyst of (S)-Binol.
Highly Enantioselective Aryl Additions of [AlAr3 (thf)] to Ketones Catalyzed by a Titanium (IV) Catalyst of (S)-Binol.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany) (2007)
Chen C-A, et al.