MilliporeSigma
  • Peloruside- and laulimalide-resistant human ovarian carcinoma cells have ?I-tubulin mutations and altered expression of ?II- and ?III-tubulin isotypes.
Peloruside- and laulimalide-resistant human ovarian carcinoma cells have ?I-tubulin mutations and altered expression of ?II- and ?III-tubulin isotypes.
Molecular Cancer Therapeutics (2011)
Kanakkanthara A