MilliporeSigma
  • TNF-a and IL10 polymorphisms interaction increases the risk of ankylosing spondylitis in Chinese Han population.
TNF-a and IL10 polymorphisms interaction increases the risk of ankylosing spondylitis in Chinese Han population.
International Journal of Clinical and Experimental Pathology (2015)
Wang NG