MilliporeSigma
Multicenter evaluation of analytical characteristics of the Elecsys
Clinical biochemistry (2016-10-12)
Stefan Palme, Robert H Christenson, Saeed A Jortani, Richard E Ostlund, Ruediger Kolm, Guido Kopal, Ruediger P Laubender
ABSTRACT
The multifunctional cytokine IL-13 is thought to play a central role in Type 2 inflammation in asthma. Serum periostin has been explored as a candidate biomarker for evaluating IL-13 activity in the airway. We describe the technical performance characteristics of a novel, fully automated immunoassay for the determination of periostin in serum. Limit of blank [LoB], limit of detection [LoD] and limit of quantitation [LoQ], linearity, precision and reproducibility across sites and lots were evaluated according to Clinical and Laboratory Standards Institute guidelines. Interferences and sample stability were also investigated. The pre-specified values for LoB (2ng/mL), LoD (4ng/mL) and LoQ (10ng/mL) were met. The assay was linear throughout the measuring range (10-160ng/mL) with recoveries within ±10% of target at concentrations >30ng/mL and within ±3ng/mL at concentrations ≤30ng/mL. Recovered periostin concentrations were also within ±10% of target in presence of 43 potentially interfering substances and drugs. Samples were stable across various storage conditions and durations (24h at room temperature, 7days at 4°C, 12weeks at -20°C, and 3 freeze/thaw cycles). Repeatability experiments resulted in CVs across samples and controls ranging from 0.9-1.5%. Intermediate precision was 1.2-1.7% and reproducibility including 3 testing sites and 3 reagent lots was 1.7-3.1%. The final assay correlates to the assay version used in previous clinical trials (Pearson's r=0.998, bias at 50ng/mL=1.2%). The performance evaluation of the Elecsys
Sigma-Aldrich
Periostin mouse, recombinant, expressed in CHO cells, FLAG® tagged, >95% (SDS-PAGE)