MilliporeSigma
  • Simulated distillation of wax samples using supercritical fluid and high temperature gas chromatography
Simulated distillation of wax samples using supercritical fluid and high temperature gas chromatography
Journal of Separation Science (1992)
Thomson.S.J and Raynaski.F.R