MilliporeSigma
  • LSD1 Inhibitor T-3775440 Inhibits SCLC Cell Proliferation by Disrupting LSD1 Interactions with SNAG Domain Proteins INSM1 and GFI1B.
LSD1 Inhibitor T-3775440 Inhibits SCLC Cell Proliferation by Disrupting LSD1 Interactions with SNAG Domain Proteins INSM1 and GFI1B.
Cancer research (2017-07-02)
Shinji Takagi, Yoshinori Ishikawa, Akio Mizutani, Shinji Iwasaki, Satoru Matsumoto, Yusuke Kamada, Toshiyuki Nomura, Kazuhide Nakamura
ABSTRACT
T-3775440 is an irreversible inhibitor of the chromatin demethylase LSD1, which exerts antiproliferative effects by disrupting the interaction between LSD1 and GFI1B, a SNAG domain transcription factor, inducing leukemia cell transdifferentiation. Here, we describe the anticancer effects and mechanism of action of T-3775440 in small-cell lung cancer (SCLC). T-3775440 inhibited proliferation of SCLC cells
Sigma-Aldrich
GSK-LSD1, ≥98% (HPLC)