- Whitefly attraction to rosemary (Rosmarinus officinialis L.) is associated with volatile composition and quantity
Whitefly attraction to rosemary (Rosmarinus officinialis L.) is associated with volatile composition and quantity
PLoS ONE (2017)
Sadeh D, et al.