MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Bioconversion of isoeugenol to vanillin and vanillic acid using the resting cells of
Bioconversion of isoeugenol to vanillin and vanillic acid using the resting cells of
3 Biotech (2017-10-06)
Morahem Ashengroph, Jahanshir Amini
ABSTRACT
40 isoeugenol-tolerant yeasts were isolated from the rhizosphere soil samples which in turn were collected from aromatic plants in different regions of Iran, and further tested for their ability to grow on a minimal medium containing isoeugenol as the sole carbon and energy source. Nine isolates which were able to grow on isoeugenol were examined for their ability to convert isoeugenol into vanillin under growing cell experiments. Of the tested yeasts, the highest conversion efficiency was observed in isolate MP24. The isolate was identified as
Sigma-Aldrich
Isoeugenol, 98%, mixture of cis and trans
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%