MilliporeSigma
  • Electrochemical Detection of Hydroxylamine via Au-Pt Alloy Nanoparticle-modified Single-walled Carbon Nanotube Electrodes.
Electrochemical Detection of Hydroxylamine via Au-Pt Alloy Nanoparticle-modified Single-walled Carbon Nanotube Electrodes.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2017-09-12)
Yanfang Geng, Euna Ko, Van-Khue Tran, Woo Sung Chung, Chan Ho Park, Min Ki Kim, Ga Hyun Jin, Gi Hun Seong
ABSTRACT
In the present study, we developed an electrochemical sensor for highly sensitive detection of hydroxylamine using Au-Pt alloy nanoparticles. Au-Pt alloy nanoparticles were electrochemically deposited on a working electrode made of single-walled carbon nanotubes. The framework composition in the Au-Pt alloy nanoparticle was easily controlled by adjusting the Au
Sigma-Aldrich
Chloroplatinic acid hexahydrate, ACS reagent, ≥37.50% Pt basis