MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Enhanced fluorescence with nanosecond dynamics in the solid state of metal ion complexes of alkoxy salophens.
Enhanced fluorescence with nanosecond dynamics in the solid state of metal ion complexes of alkoxy salophens.
Physical chemistry chemical physics : PCCP (2017-11-04)
Tuhin Khan, Anindya Datta
ABSTRACT
Salophen (N,N'-bis-(salicylidene)-o-phenylenediamine) is a Schiff base, which is weakly emissive in solution, but strongly emissive in the solid state. The trend is reversed in its aluminium complex (SalAl
Sigma-Aldrich
2,3-Dihydroxybenzaldehyde, 97%
Sigma-Aldrich
o-Vanillin, 99%
Sigma-Aldrich
3-Ethoxysalicylaldehyde, 97%