MilliporeSigma
  • Leptin receptor polymorphism is associated with serum lipid levels and impairment of cholesterol lowering effect by simvastatin in Japanese men.
Leptin receptor polymorphism is associated with serum lipid levels and impairment of cholesterol lowering effect by simvastatin in Japanese men.
Diabetes research and clinical practice (2003-11-20)
Akiko Takahashi-Yasuno, Hiroaki Masuzaki, Takashi Miyawaki, Yoshihiro Ogawa, Naoki Matsuoka, Tatsuya Hayashi, Kiminori Hosoda, Gen Inoue, Yasunao Yoshimasa, Kazuwa Nakao
ABSTRACT
To investigate whether leptin receptor (Ob-R) Arg223Gln polymorphism influences serum lipid levels and whether this polymorphism affects the efficiency of the cholesterol lowering HMG-CoA reductase inhibitor, simvastatin [Clin. Cardiol. 16 (1993) 317]. Case-control association study. We studied 201 Japanese men without medical care, and 78 Japanese who took simvastatin. Genotyping was performed by polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP). Serum lipid and leptin levels were determined. Subjects with the Arg/Arg homozygotes had significantly higher serum total cholesterol (TC) and low density lipoprotein cholesterol (LDL-C) levels than those with the Arg/Gln heterozygotes and Gln/Gln homozygotes (TC: Arg/Arg: 213+/-3, Arg/Gln: 196+/-6, Gln/Gln: 184+/-5, P=0.004 for comparison among three genotypes, P=0.008 for difference between Arg/Arg and Arg/Gln, and P=0.025 for difference between Arg/Arg and Gln/Gln, LDL-C: Arg/Arg: 127+/-3, Arg/Gln: 112+/-6, Gln/Gln: 114+/-8, P=0.027) for comparison among three genotypes and P=0.011 for difference between Arg/Arg and Arg/Gln. Subjects with the Arg/Arg homozygotes had significantly lower serum high density lipoprotein cholesterol (HDL-C) levels than those with the Arg/Gln heterozygotes and Gln/Gln homozygotes (Arg/Arg: 55+/-1, Arg/Gln: 62+/-3, Gln/Gln: 57+/-7, P=0.046) for comparison among three genotypes and P=0.013 for difference between Arg/Arg and Arg/Gln. In addition, in 78 patients with hypercholesterolemia who took 5 mg simvastatin, the TC lowering effect by simvastatin in subjects with the Arg/Arg homozygotes was significantly lower than in those with the Arg/Gln heterozygotes and Gln/Gln homozygotes (the reduction in serum TC levels; 62+/-4 vs. 79+/-6, P=0.044). We demonstrate that Ob-R Arg223Gln polymorphism in Japanese men is associated with significant elevation of serum TC and LDL-C levels. Our data also show that the Arg/Arg homozygotes tend to show lowered level of serum HDL-C. Furthermore, this polymorphism tends to show an attenuated response to an HMG-CoA reductase inhibitor in terms of the cholesterol lowering effect. These results suggest that the Ob-R gene may serve as a novel modifier gene for hypercholesterolemia in Japanese men.