MilliporeSigma
  • A transcription factor involved in skeletal muscle gene expression is deleted in patients with Williams syndrome.
A transcription factor involved in skeletal muscle gene expression is deleted in patients with Williams syndrome.
European journal of human genetics : EJHG (1999-11-26)
M Tassabehji, M Carette, C Wilmot, D Donnai, A P Read, K Metcalfe
ABSTRACT
Williams-Beuren syndrome (WS) is a developmental disorder caused by a hemizygous microdeletion of approximately 1.4MB at chromosomal location 7q11.23. The transcription map of the WS critical region is not yet complete. We have isolated and characterised a 3.4 kb gene, GTF3, which occupies about 140 kb of the deleted region. Northern blot analysis showed that the gene is expressed in skeletal muscle and heart, and RT-PCR analysis showed expression in a range of adult tissues with stronger expression in foetal tissues. Part of the conceptual GTF3 protein sequence is almost identical to a recently reported slow muscle-fibre enhancer binding protein MusTRD1, and shows significant homology to the 90 amino-acid putative helix-loop-helix repeat (HLH) domains of the transcription factor TFII-I (encoded for by the gene GTF2I). These genes may be members of a new family of transcription factors containing this HLH-like repeated motif. Both GTF3 and GTF2I map within the WS deleted region, with GTF2I being positioned distal to GTF3. GTF3 is deleted in patients with classic WS, but not in patients we have studied with partial deletions of the WS critical region who have only supravalvular aortic stenosis. A feature of WS is abnormal muscle fatiguability, and we suggest that haploinsufficiency of the GTF3 gene may be the cause of this.