MilliporeSigma
Sequence and functional characterization of feline interleukin 2.
Biochemical and biophysical research communications (1993-08-16)
P J Cozzi, P A Padrid, J Takeda, M L Alegre, N Yuhki, A R Leff
ABSTRACT
Since the discovery of its involvement in the pathogenesis of feline immunodeficiency virus infection ("cat AIDS") and feline leukemia virus infection, the role of feline interleukin 2 (IL-2) has been a focus of particular interest. The purpose of this study was to clone feline IL-2 cDNA, as well as synthesize bioactive recombinant feline IL-2. The isolation of cDNA encoding feline IL-2 was carried out using a PCR-based strategy and screening of a feline leukocyte cDNA library. Feline IL-2 consists of 154 amino acids including a putative signal sequence and has 81%, 69%, 60% and 64% identity to human, bovine, murine and rat IL-2, respectively. Feline IL-2 cDNA was expressed in COS-7 cells. The secreted protein has CTLL-4 murine cytotoxic T cell proliferative activity characteristic of authentic IL-2. These data confirm the synthesis of bioactive recombinant feline IL-2.