MilliporeSigma
  • The influence of cyclooxygenase-1 expression on the efficacy of cyclooxygenase-2 inhibition in head and neck squamous cell carcinoma cell lines.
The influence of cyclooxygenase-1 expression on the efficacy of cyclooxygenase-2 inhibition in head and neck squamous cell carcinoma cell lines.
Anti-cancer drugs (2011-02-09)
Seok-Woo Park, Hyo-Sun Kim, Myung-Sun Choi, Ji-Eun Kim, Woo-Jin Jeong, Dae-Seog Heo, Myung-Whun Sung
ABSTRACT
We have previously observed that cyclooxygenase-2 (COX-2) inhibition blocked the production of vascular endothelial growth factor (VEGF) in some head and neck squamous cell carcinoma (HNSCC) cells. However, as some HNSCC cells showed little response to COX-2 inhibition, although they highly expressed COX-2 and prostaglandin E2, we set out to elucidate what made this difference between them and focused on the possibility of the differential expression of COX-1. In western blotting, we found that COX-1 was expressed in SNU-1041 and SNU-1066, but not in SNU-1076 and PCI-50. Only in those cell lines without expression of COX-1 was VEGF production blocked meaningfully by small interfering RNA of COX-2. However, by cotreating with small interfering RNAs of COX-2 and COX-1, VEGF synthesis and prostaglandin E2 were inhibited in SNU-1041 and SNU-1066, similarly in SNU-1076 and PCI-50 with high expression of only COX-2. We also found that there was no difference in the pattern of prostaglandin synthesis between COX-2 and COX-1 through enzyme-linked immunosorbent assay for various prostaglandins. Our study suggests that, as COX-1 and COX-2 express and affect VEGF synthesis in HNSCC cells, we should check COX-1 expression in investigations on cancer treatment by inhibiting COX-2-induced prostaglandins.