MilliporeSigma
AGT and AT1R gene polymorphism in hypertensive heart disease.
International journal of clinical practice (2002-11-12)
M Mettimano, V Romano-Spica, A Ianni, Ml Specchia, A Migneco, L Savi
ABSTRACT
Left ventricular hypertrophy in patients with hypertension is a main clinical prognostic entity The aim of this study was to evaluate the association between mutations at genes of the renin-angiotensin system (RAS) and the development of left ventricular hypertrophy. Genetic polymorphism in angiotensinogen (AGT) and angiotensin Il-type 1 receptor (AT1R) genes was examined in a group of well-selected essential hypertensive caucasians with left ventricular involvement (n = 40) and a group of healthy unrelated caucasians (n = 150). Cardiac morphology and function were assessed by M-mode echocardiography. Molecular variants were analysed by amplified fragment length polymorphism. We observed a statistically significant difference both for AGT and AT1R genotype distribution in patients with left ventricular hypertrophy compared with controls (p<0.05). A 0.49 and 0.225 frequency was detected among cases for T and C mutant alleles at AGT and AT1R genes. Mutations in RAS genes are involved in the pathophysiology of target-organ damage in essential hypertension. Evaluation of molecular factors conferring a risk of developing heart involvement may lead to better identification of patient subgroups and more effective control of the clinical course.