MilliporeSigma
  • Analysis of interaction partners for eukaryotic translation elongation factor 1A M-domain by functional proteomics.
Analysis of interaction partners for eukaryotic translation elongation factor 1A M-domain by functional proteomics.
Biochimie (2011-06-22)
Annalisa Lamberti, Carmen Sanges, Angela Chambery, Nunzia Migliaccio, Francesco Rosso, Antimo Di Maro, Ferdinando Papale, Monica Marra, Augusto Parente, Michele Caraglia, Alberto Abbruzzese, Paolo Arcari
ABSTRACT
The eukaryotic translation elongation factor 1A (eEF1A), besides to its canonical role in protein synthesis, is also involved in several other cellular processes, depending on changes in cellular location, cell type, concentration of ligands, substrates or cofactors. Therefore eEF1A is a moonlighting protein that participates to a network of molecular interactions involving its structural domains. Since the identification of novel protein-protein interactions represents important tasks in post-genomic era, the interactome of eEF1A1 M-domain was investigated by using a proteomic approach. To this purpose, the eEF1A1 M-domain was fused with glutathione-S-transferase (GST) and Strep-tag (ST) at it's N- and C-terminal, respectively. The recombinant protein (GST-M-ST) was purified and incubated with a mouse embryo lysate by applying an affinity chromatography strategy. The interacting proteins were separated by SDS-PAGE and identified by peptide mass fingerprinting using MALDI-TOF mass spectrometry. Besides the known partners, the pool of interacting proteins contained sorbin, a polypeptide of 153 amino acids present in SH3 domain-containing adaptor proteins, such as SORBS2. This interaction was also assessed by Western blot on immunoprecipitate from mouse embryo or H1355 cell lysates with anti-eEF1A or anti-SORBS2 antibodies and on eEF1A1-His pull-down from H1355 cell lysate with antibody anti-SORBS2. Furthermore, the interaction between eEF1A and SORBS2 was also confirmed by confocal microscopy and FRET analysis. Interestingly, a co-localization of SORBS2 and eEF1A was evidenced at level of plasma membrane, thus suggesting the involvement of eEF1A1 in novel key signal transduction complexes.