MilliporeSigma
HPLC Analysis of L-Cysteine on SeQuant® ZIC-HILIC
HPLC Analysis of L-Cysteine on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 μm, 200 Å (1.50453.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate
0.5 mL/min
column temp.
ambient
detector
220 nm
injection
1 μL
sample
1 mg/mL of acetylcysteine in acetonitrile/water (70/30)
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany