Growth Direct® System
Partnering to Deliver Automated Solutions for Your QC Lab
While we are designing new solutions internally to automate QC workflows, we also seize every opportunity to partner with leading-edge companies. We signed a distribution and collaboration agreement with the company Rapid Micro BioSystems, Inc. This partnership allows us to add to our portfolio the Growth Direct® system, a fully automated microbial quality control solution, reinforcing our commitment to innovation in pharmaceutical manufacturing and microbiology.
Automated Microbial Detection and Enumeration Platform
Accelerate quality control processes with the fully automated Growth Direct® system.* This innovative platform improves data integrity, reducing human error, and streamlines microbiological testing across your laboratory.
The Growth Direct® system* is a fully automated, non-destructive platform designed for microbial quality control (QC) testing in modern laboratories. It replaces outdated manual and paper-based methods with a digital, automated approach, delivering faster results and improved compliance for various testing needs.
Whether performing bioburden testing, environmental monitoring, or accelerating sterility test results, the Growth Direct® system* provides rapid and accurate results while minimizing human error and enhancing data integrity.
Key features include:
- Automated colony enumeration and detection
- Touch-free incubation and imaging
- Seamless integration with LIMS and digital QC workflows
- 21 CFR Part 11–compliant electronic records
- Non-destructive testing method allows for downstream analysis
Discover how the Growth Direct® system* can transform your microbial quality control processes.
Enhanced Compliance and Data Integrity
Eliminate transcription errors and manual recordkeeping with fully digital data management and audit-ready reports.
Operational Efficiency
Free up analyst time and standardize results across shifts and sites with trusted automation.
Click here to learn more about other automated solutions we are developing internally in partnership with users and engineering companies.
Related Resources
- Data Sheet: Automate Your Bioburden, Environmental Monitoring and Water Testing
Understand more about the benefits of using the Growth Direct® system.
- Data Sheet: The Growth Direct® System, Purposely Designed for Regulated Microbiology Demands
Explore the complete range of outstanding features the system offers.
- Flyer: Mold Detection in Real Time Thanks to the RMBNucleus™ Mold Alarm
Get details about a key functionality of the Growth Direct® system, enabling automated alerts triggered by detection of molds.
Join our experts to learn what to consider when evaluating and implementing automated rapid solutions.
*Growth Direct® is a registered trademark of Rapid Micro Biosystems, Inc