MilliSentials™ Essential Lab Equipment for Productive, Efficient Labs
Work Smarter
Many protocols require general lab equipment (vortexers, pipette controllers) to tackle routine, repetitive tasks in the laboratory. Unfortunately, current offerings for this equipment are characterized by decades old designs lacking any recent innovation.
We are proud to introduce the MilliSentials™ line of general lab equipment aimed at increasing lab efficiency through updated, innovative general lab equipment and devices. Featuring modernized, enhanced devices that simplify routine tasks in the laboratory, these products help streamline your applications.
MilliSentials™ Lab Labeling System
Adhering to your Highest Standards
MilliSentials™ Lab Labeling System for durable, chemically-resistant labels for samples and specimens
The MilliSentials™ Lab Labeling System provides a complete laboratory labeling solution with laboratory grade labels, a compact WIFI capable printer, and custom-designed laboratory labeling software.
Key Features:
  • Laboratory grade labels
    • Labels can withstand temperatures from -196 °C (liquid nitrogen) to 100 °C.
    • Labels provide chemical resistance to water, ethanol, IPA, DMSO, methanol, acetonitrile, bleach, acetone, xylene, and toluene
    • Adaptable labels can accommodate multiple vessel types and sizes
  • Intuitive, easy-to-use thermal transfer printer with hardwire and wireless connection options
  • Custom built laboratory software featuring pre-defined labels for laboratory vessels
    • Spreadsheet printing functionality
    • Build, save, and share label templates for common label setups
    • Access and print labels using a laptop, tablet, or mobile device on the
      same network as the printer
Product Page
MilliSentials™ Aliquoting Pipette Controller
Enhance your Liquid Handling
MilliSentials™ Aliquoting Pipette Controller for precision aliquoting
The MilliSentials™ Aliquoting Pipette Controller combines the automatic aliquoting function of a repeat pipettor with the basic functions of a standard pipette controller in a single device.
Key Features
  • Built in aliquoting capability automatically dispenses accurate aliquots between 0.5-5.0 mL at the touch of a button
  • Touch sensitive aspirate and dispense buttons allow precise control
  • Ergonomic design reduces hand strain
Product Page
MilliSentials™ Vortexer
Modernize your Vortexer
MilliSentials™ Vortexer
The MilliSentials™ Vortexer provides powerful agitation to liquid samples within sealed containers. The compact and powerful design will accommodate tubes with volumes from 0.2 to 50 mL, vials with volumes from 2.0 to 10 mL, and square bottles up to 150 mL.
The MilliSentials™ Vortexer offers powerful agitation in a compact modernized design.
Key Features:
  • Powerful agitation at a reasonable price provides a valuable option
  • Modernized compact design minimizes loss of bench space
Product Page
Related Links