MilliporeSigma
HomeServicesBioReliance® Contract TestingBulk Harvest Release TestingCell & Gene Therapy Bulk Harvest Release Testing
Cell & Gene Therapy Bulk Harvest Release Testing
Bulk lot release testing is critical to unlock the potential of new gene therapies and gene-modified cell therapies
To unlock the potential of gene therapies and gene-modified cell therapies, our testing services often play a critical role in helping biomanufacturers ensure the identity, potency, and/or safety of the unpurified bulk, raw materials, cell banks, virus banks, and plasmids that are used in production of their final products.
We offer cGMP-compliant bulk harvest release testing support for cell and gene therapies, including those that utilize:
Illustration of adeno-associated vector
AAV
Illustration of lentivirus
Retrovirus/ Lentivirus
Illustration of adenovirus
Adenovirus
Gene Therapy Testing for Unprocessed and Purified Bulk Harvest
Testing of unpurified bulk, which may include transfected producer cells, should include verification of identity, and may involve confirmation of the gene of interest, determination of viral titer, and confirmation of the absence of replication competent virus. Tests for microorganisms (e.g., bacteria, fungi, and mycoplasma) are appropriate for this material, as are broad specificity assays for adventitious viruses.
 Unprocessed Bulk HarvestPurified Bulk Harvest
Adventitious Agents✓✓
Identity✓✓
Titer✓✓
Biological Activity ✓
Residuals ✓
Replication-competent Viral Testing✓✓
Gene Expression ✓
Plasmid Monitoring ✓
Our bulk harvest biosafety and characterization testing packages include:
  • Sterility
  • Mycoplasma detection
  • Adventitious virus and virology testing, including quantification of retrovirus-like particles
  • Identity
  • Infectious titer
  • Empty/full capsid analysis
  • Replication-competent virus
In addition, our experts can help bring your therapy to the clinic and beyond with regulatory and technical support; advice on regulatory strategies; data review and evaluation; report writing and technical support; as well as responses to regulatory authorities.
To draw on our experience, use our reply form below and get in touch today.
Request Information