Technisches Datenblatt
Produktname
Titan(IV)isopropoxid ≥97.0%
Produktnummer
87560
Produktmarke
ALDRICH
CAS-Nummer
546-68-9
Molekulargewicht
284.22
Test | Spezifikation |
|---|---|
| APPEARANCE (COLOR) | COLORLESS TO FAINT YELLOW (MAY DISCOLOR ON STORAGE) |
| APPEARANCE (FORM) | CLEAR LIQUID OR SOLID MASS |
| ASSAY (GRAVIMETR.) | 97.0 - 103.0 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |