Product Name
Digitonin Used as non-ionic detergent
Product Number
D141
Product Brand
SIGMA
CAS Number
11024-24-1
Molecular Weight
1229.31
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | White to Off White |
| APPEARANCE (FORM) | Powder |
| PURITY (TLC AREA %) | APPROX. 50 % |
| SPECIFIC ROTATION (20/D) | -57 - -49 Degrees |
| CONCENTRATION | C=1 IN ETHANOL |
| SOLUBILITY (TURBIDITY) | 1 G CAN BE DISSOLVED IN 20 ML OF WATER BY HEATING TO 95 - 98 DEG C |
| WATER | ≤ 6.0 % |
| INFRARED SPECTRUM | CONFORMS TO STRUCTURE |