Product Name
Phosphate buffered saline pH 7.4, liquid, sterile-filtered, suitable for cell culture
Product Number
P4474
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH @ 25 °C7.2 - 7.6
Osmolality275 - 304 mOs/kg
Salt Toxicity TestPass
Cell LineCell Line - Cell Types
Sterility by current EP/USPPass
Endotoxin<= 1.0 EU/ml
Animal Component FreeConforms