MilliporeSigma
HomeSupportCalculators & AppsSolubility Table for Water at Temperature
Solubility Table for Water at Temperature
Water temperature can have a significant effect on the solubility of compounds. Refer to the chart below to find reference values per gram of common compounds and salts (with chemical formula) at six temperatures of 100 g of water from 0 degrees to 100 degrees Celsius. The content and density of the total solution at 20 degrees are also provided. This table complements our Solubility Rules & Chart and Miscibility Table.
Solubility of inorganic compounds in water in relation to temperature
Compound NameChemical FormulaSolubility in g/100 g H2O at °CContent
of the total solution at
20 °C in %
Density
of the total solution
at
20 °C in %
020406080100
Aluminum ammonium sulfate dodecahydrateAlNH4(SO4)2 · 12H2O2.66.5912.3621.135.2109.2
(95 °C)
6.181.0459
(15.5 °C)
Aluminum chloride hexahydrateAlCl3 · 6H2O44.945.646.34747.7–31.3–
Aluminum nitrate nonahydrateAl(NO3)3 · 9H2O6175.4489108––43–
Aluminum potassium sulfate dodecahydrateAlK(SO4)2 · 12H2O2.966.0113.633.372109
(90 °C)
5.671.053
Aluminum sulfate octadecahydrateAl2(SO4)3 · 18H2O31.236.4445.658738926.71.308
Ammonium bromideNH4Br60.675.591.1107.8126.7145.643.9–
Ammonium chlorideNH4Cl29.737.564655.365.677.327.31.075
Ammonium dihydrogen phosphateNH4H2PO422.736.856.782.9120.717426.9–
Ammonium hydrogen carbonateNH4HCO311.921.2236.659.2109.235517.51.07
di-Ammonium hydrogen phosphate(NH4)2HPO457.568.681.897.6-115.5–40.71.3436
(14.5 °C)
Ammonium iron(II) sulfate hexahydrate(NH4)2Fe(SO4)2 · 6H2O17.826.938.553.473–21.21.18
Ammonium mono-
vanadate
NH4VO3–4.813.2–––––
Ammonium nitrateNH4NO3118.5187.72834156101000651.308
Ammonium sulfate(NH4)2SO470.475.4481.287.494.1102431.247
Ammonium thiocyanateNH4SCN115163235347-525–61.98–
Antimony(III) chlorideSbCl3601.6931.513684531––90.31–
Barium acetateBa(CH3COO)2587279747474––
Barium chloride dihydrateBaCl2 · 2H2O30.735.740.846.452.558.726.31.28
Barium hydroxide octahydrateBa(OH)2 · 8H2O1.53.488.221––3.361.04
Barium nitrateBa(NO3)24.959.0614.420.327.234.28.31.0691
di-Boron trioxideB2O31.12.246.29.515.72.15–
Boric acidH3BO32.665.0428.714.823.639.74.81.015
Cadmium sulfate octahydrate3CdSO4 · 8H2O75.7576.6979.2681.984.6–43.41.616
Calcium chloride dihydrateCaCl2 · 2H2O––128.1136.8147159––
Calcium nitrate tetrahydrateCa(NO3)2 · 4H2O101129.39196–––56.4–
Calcium sulfate dihydrateCaSO4 · 2H2O0.1760.360.21220.20470.19660.16190.2041.001
Cesium chlorideCsCl161187208230250271––
Cesium nitrateCsNO39.32347.283.8134197––
Chromium(VI) oxideCrO3163166.7217117618919962.51.71
(16.5 °C)
Cobalt chloride hexahydrateCoCl2 · 6H2O83.5100126169.5 (56 °C)   
Cobalt nitrate
hexahydrate
Co(NO3)2 · 6H2O––50–    
Cobalt sulfate heptahydrateCoSO4 · 7H2O25.536.2649.9–––26.6–
Copper(I) chlorideCuCl–1.52
(25 °C)
––––1.497
(25 °C)
–
Copper(II) chloride dihydrateCuCl2 · 2H2O70.657783.891.299.2107.943.51
Copper(II) nitrate trihydrateCu(NO3)2 · 3H2O––160179208-257––
Copper (II) sulfate anhydrousCuSO425.536.248607083––
Copper(II) sulfate pentahydrateCuSO4 · 5H2O14.820.772939.153.673.617.21.1965
Iron(III) chloride hexahydrateFeCl3 · 6H2O74.591.94––––47.91.52
Iron(II) chloride tetrahydrateFeCl2 · 4H2O–62.3568.678.3––38.41.49
Iron(II) sulfate heptahydrateFeSO4 · 7H2O15.6526.5840.347.6
(50 °C)
––211.225
Iron(II) sulfate monohydrateFeSO4 · H2O––––43.8-31.6––
Iron (III) chloride hexahydrateFeCl3 · 6H2O74.591.94––––47.91.52
Lead chloridePbCl20.67280.991.451.982.623.310.981.007
Lead nitratePb(NO3)236.452.2269.488107.5127.334.31.4
Lithium carbonateLi2CO3–1.33––––1.31–
Lithium chloride monohydrateLiCl · H2O–82.8290.4100113-127.545.31.29
Lithium sulfate monohydrateLiSO4 · H2O36.234.833.532.331.53125.61.23
Magnesium chloride hexahydrateMgCl2 · 6H2O52.854.5757.560.765.8772.735.31.331
Magnesium nitrate hexahydrateMg(NO3)2 · 6H2O63.970.0781.893.7––41.21.388
(25 °C)
Magnesium sulfate heptahydrateMgSO4 · 7H2O30.05
(10 °C)
35.645.4–––26.251.31
Manganese(II) chloride tetrahydrateMnCl2 · 4H2O63.673.6288.7-106
(58.1 °C)
––42.41.499
Manganese(II) chloride dihydrateMnCl2 · 2H2O––––110.5115––
Manganese(II) sulfate monohydrateMnSO4 · H2O––6058.645.535.5––
Mercury(II) bromideHgBr2–0.62
(25 °C)
-0.961.672.774.90.62
(25 °C)
–
Mercury(II) chlorideHgCl24.296.619.613.924.254.16.21.052
Nickel chloride
hexahydrate
NiCl2 · 6H2O51.755.3––––35.61.46
Nickel nitrate hexahydrateNi(NO3)2 · 6H2O79.294.1118.8–––48.5–
Nickel sulfate hexahydrateNi2SO4 · 6H2O–––57––––
Potassium bromateKBrO33.16.813.12233.949.76.41.048
Potassium bromideKBr5465.8576.185.995.3104.939.71.37
Potassium carbonateK2CO3105.5111.511712714015652.81.58
Potassium chlorateKClO33.37.314.525.939.756.26.81.042
Potassium chlorideKCl28.1534.2440.345.65156.225.51.174
Potassium chromateK2CrO45963.686770.975.179.238.91.378
Potassium cyanideKCN-6371.6
(25 °C)
–81
(50 °C)
-95
(75 °C)
122
(103.3 °C)
41.73
(25 °C)
–
Potassium dichromateK2Cr2O74.6812.4926.345.67310311.11.077
Potassium dihydrogen phosphateKH2PO414.322.733.948.668–18.5–
Potassium disulfiteK2S2O527.544.963.98510813330.99–
Potassium hexachloroplatinate(IV)K2 [Pt(Cl)6]0.741.11.72.63.85.2––
Potassium hexacyanoferrate(II) trihydrateK4 [Fe(CN)6] · 3H2O1528.8742.75668.9-82.722.41.16
Potassium hexacyanoferrate(III)K3 [Fe(CN)6]29.94659.570.981.891.631.511.18
Potassium hydrogen carbonateKHCO322.633.345.360––24.981.18
di-Potassium hydrogen phosphate trihydrateK2HPO4 · 3H2O–159212.5–––61.39–
di-Potassium hydrogen phosphate anhydrousK2HPO4–––266––––
Potassium hydrogen sulfateKHSO436.351.476.3––121.633.95–
Potassium hydroxide monohydrateKOH · H2O––136.4147160178––
Potassium iodateKlO34.78.1112.918.524.832.37.51.064
Potassium iodideKl127.8144.51161176.2191.520859.11.71
Potassium nitrateKNO313.2531.6663.9109.9169245.224.11.16
di-Potassium oxalate monohydrateK2C2O4 · H2O–35.88––––26.4–
Potassium perchlorateKClO40.761.733.637.1813.3822.21.71.008
Potassium permanganateKMnO42.836.4312.5622.4––61.04
Potassium peroxodisulfateK2S2O80.180.471.1–––0.468–
Potassium sulfateK2SO47.3311.1114.7918.221.2924.1101.0807
Potassium thiocyanateKSCN177218––––68.551.42
Rubidium chlorideRbCl70.683.6–––128––
Silver nitrateAgNO3115219.2334.8471652102468.62.18
Silver sulfateAg2SO40.5730.7960.9791.151.31.460.75–
Sodium acetate trihydrateNaCH3COO · 3H2O36.346.4265.4138
(58 °C)
––31.71.17
Sodium bromideNaBr–––118118.3121.2––
Sodium carbonate decahydrateNa2CO3 · 10H2O6.8621.66––––17.81.1941
Sodium carbonate monohydrateNa2CO3 · H2O––48.946.244.544.5––
Sodium carbonate anhydrousNa2CO37.121.448.546.545.845.5––
Sodium chlorateNaClO380.598.82115.2-138-16720449.7–
Sodium chlorideNaCl35.635.836.4237.0538.0539.226.41.201
Sodium dichromate dihydrateNa2Cr2O7 · 2H2O163.2180.16220.5283385–64.3–
Sodium dihydrogen phosphate dihydrateNaH2PO4 · 2H2O57.785.2138.2–––46–
Sodium dihydrogen phosphate anhydrousNaH2PO4–––179.3207.3284.4––
tetra-Sodium diphosphate decahydrateNa4P2O7 · 10H2O2.75.4812.521.93040.265.21.05
Sodium disulfiteNa2S2O5–65.371.179.988.7-10039.5–
Sodium fluorideNaF-3.64.1––––3.941.04
Sodium hydrogen carbonateNaHCO36.899.612.71619.723.68.761.08
di-Sodium hydrogen phosphate dodecahydrateNa2HPO4 · 12H2O1.637.7––––7.21.08
di-Sodium hydrogen phosphate heptahydrateNa2HPO4 · 7H2O––55–––––
di-Sodium hydrogen phosphate dihydrateNa2HPO4 · 2H2O–––8392.4–––
di-Sodium hydrogen phosphate anhydrousNa2HPO4–––––104.1––
Sodium hydroxide monohydrateNaOH · H2O–109.22126178––52.21.55
Sodium hydroxideNaOH––––313.7341––
Sodium iodateNalO3––––2732.8––
Sodium iodideNal––––295303––
Sodium nitrateNaNO370.788.3104.9124.714817646.81.38
Sodium nitriteNaNO27384.5295.7112.3135.516345.81.33
Sodium perchlorate monohydrateNaClO4 · H2O167181243–––64.411.757
(25 °C)
tri-Sodium phosphate dodecahydrateNa3PO4 · 12H2O1.512.1131558110810.81.106
Sodium sulfate decahydrateNa2SO4 · 10H2O4.5619.19––––16.11.15
Sodium sulfate anhydrousNa2SO4––48.145.2643.0942.3––
Sodium sulfiteNa2SO3––3733.22926.6––
di-Sodium tetraborateNa2B4O71.22.7620.331.552.5––
Sodium thiosulfate pentahydrateNa2S2O3 · 5H2O52.570.07102.6–––41.21.39
Sodium thiosulfate anhydrousNa2S2O3––––245266––
Strontium chloride hexahydrateSrCl2 · 6H2O44.153.8566.685.2––351.39
Strontium hydroxide octahydrateSr(OH)2 · 8H2O0.350.71.53.137.0224.20.69–
Strontium nitrateSr(NO3)2––91.294.297.2101.2––
Zinc chlorideZnCl2––453488541614––
Zinc nitrate tetrahydrateZn(NO3)2 · 4H2O––211.5–––––
Zinc sulfate heptahydrateZnSO4 · 7H2O41.653.8––––351.47
Zinc sulfate monohydrateZnSO4 · H2O–––76.566.760.5––