MilliporeSigma
  • Impact of 2-Vinylpyridine as Electrolyte Additive on Surface and Electrochemistry of Graphite for C/ LiMn2O4 Li-Ion Cells
Impact of 2-Vinylpyridine as Electrolyte Additive on Surface and Electrochemistry of Graphite for C/ LiMn2O4 Li-Ion Cells
Journal of the Electrochemical Society (2005)
Komaba S, et al.