- Gas-phase synthesis of [Ag4H]+ and its mediation of the C-C coupling of allyl bromide
Gas-phase synthesis of [Ag4H]+ and its mediation of the C-C coupling of allyl bromide
Angewandte Chemie (International Edition in English) (2005)
Khairallah GN and O'Hair, Richard AJ