MilliporeSigma
  • Preparation and properties of novel double-chain nonionic surfactants with acid decomposition function.
Preparation and properties of novel double-chain nonionic surfactants with acid decomposition function.
Journal of oleo science (2010-06-02)
Daisuke Ono, Hirofumi Sato, Motohiro Shizuma, Masaki Nakamura
ABSTRACT
Novel double-chain nonionic surfactants with an acid decomposition function were prepared by acid-catalyzed condensation of chloroacetone with fatty alcohols (octyl, decyl, and dodecyl), followed by a Williamson reaction with polyethylene glycol without any expensive reagents and special equipment. These surfactants showed easy micelle formation compared to those of polyoxyethylene (n=9) dodecyl ether (C(12)EO9), and good foaming properties. The emulsion stability of these surfactants was almost the same as that of C(12)EO9. They decomposed completely after 30 min at pH 1. After 28 days they were more than 60% biodegradable and were almost the same as sodium dodecanoate.
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Chloroacetone, Wacker Chemie AG, ≥96.0% (GC)
Sigma-Aldrich
Chloroacetone, 95%